missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, TRC

Product Code. 30011637
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011637 100 g 100g
30006950 10 g 10g
30007896 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011637 Supplier Toronto Research Chemicals Supplier No. TRCP755995100G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5 H9 N O2
Molecular Weight (g/mol) 115.13
Synonym Proline, (S)-pyrrolidine-2-carboxylic acid, (-)-2-Pyrrolidinecarboxylic acid, (-)-Proline, (S)-(-)-Proline, (S)-2-Carboxypyrrolidine, (S)-2-Pyrrolidinecarboxylic acid, (S)-Proline, 2-Pyrrolidinecarboxylic acid, Carboxypyrrolidine, L-(-)-Proline, L-Pyrrolidine-2-carboxylic acid, L-α-Pyrrolidinecarboxylic acid, NSC 46703
IUPAC Name (2S)-pyrrolidine-2-carboxylic acid
SMILES OC(=O)[C@@H]1CCCN1

Specifications

InChI Formula InChI=1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1
Quantity 100 g
Formula Weight 115.0633
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Proline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.