missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11398836
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398836 1 g 1g
11308846 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398836 Supplier Thermo Scientific Alfa Aesar Supplier No. L19489.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5497-76-7
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00153459
InChI Key IUUYANMOEMBTBV-FJXQXJEOSA-N
Synonym h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl
PubChem CID 6453107
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES CC(C)(C)OC(=O)C1CCCN1.Cl

Specifications

Melting Point 110°C to 112°C
Beilstein 3596764
Quantity 1 g
Formula Weight 207.7
Optical Rotation −31° (c=0.5 in Water)
Percent Purity ≥98%
Chemical Name or Material L-Proline tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.