missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11308846
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11308846

Brand: Thermo Scientific Alfa Aesar L19489.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5497-76-7
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00153459
InChI Key IUUYANMOEMBTBV-FJXQXJEOSA-N
Synonym h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl
PubChem CID 6453107
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES CC(C)(C)OC(=O)C1CCCN1.Cl

Specifications

Melting Point 110°C to 112°C
Beilstein 3596764
Quantity 5 g
Formula Weight 207.7
Optical Rotation −31° (c=0.5 in Water)
Percent Purity ≥98%
Chemical Name or Material L-Proline tert-butyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.