missing translation for 'onlineSavingsMsg'
Learn More

L-Proline benzyl ester hydrochloride, 98%, ACROS Organics™

Product Code. 10723904 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10723904

Brand: Thermo Scientific Acros 320600010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16652-71-4
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
MDL Number MFCD00039002
InChI Key NEDMOHHWRPHBAL-MERQFXBCSA-N
Synonym l-proline benzyl ester hydrochloride, h-pro-obzl.hcl, l-proline benzyl ester hcl, s-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl l-prolinate hydrochloride, proline, phenylmethyl ester, hydrochloride, proline benzyl ester hydrochloride, benzyl 2s-pyrrolidine-2-carboxylate hydrochloride, l-proline, phenylmethyl ester, hydrochloride, phenylmethyl 2s pyrrolidine-2-carboxylate, chloride
PubChem CID 2733413
IUPAC Name benzyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl

Specifications

Melting Point 148.0°C to 151.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4365
Quantity 1 g
Specific Rotation Condition −44° (20°C c=1,methanol)
Specific Rotation − 44.00
Solubility Information Solubility in water: miscible. Other solubilities: soluble in acetone, benzene, ethyl ether, ethanol
Formula Weight 241.72
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Proline benzyl ester hydrochloride, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.