missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, >99%, MP Biomedicals™

Product Code. 11496082 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496082 100 g 100g
11406092 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496082 Supplier MP Biomedicals Supplier No. 0210273080
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 147-85-3 | C5H9NO2 | 115.13 g/mol

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
PubChem CID 145742
ChEBI CHEBI:17203
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

Color White
Assay Percent Range >99%
Quantity 100 g
Formula Weight 115.13
Percent Purity >99%
Grade Cell Culture
Physical Form Powder
Chemical Name or Material L-Proline, Cell Biology and Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.