missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
Description
- L-Proline is used in cell culture media.
- L-Proline is also a component of MEM non-essential amino acids solution.
Specifications
Specifications
| Melting Point | 221°C |
| Solubility Information | Soluble in water, 162,000 mg/L at 25°C |
| Density | 1.064 g/mL at 24°C |
| Quantity | 100 g |
| Formula Weight | 115.1 |
| Optical Rotation | -82° to -85° (c=1.6, water) |
| Percent Purity | ≥99% |
| Grade | Cell Culture |
| Physical Form | Powder |
| pKa | 1.95 (COOH), 10.64 (NH2) |
| Show More |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction