missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, 99%

Product Code. 11449512
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449512 100 g 100g
11439512 25 g 25g
11459512 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449512 Supplier Thermo Scientific Alfa Aesar Supplier No. A10199.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 147-85-3 | C5H9NO2 | 115.13 g/mol

L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.

Solubility
Soluble in water and ethanol. Insoluble in ether, butanol and isopropanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
PubChem CID 145742
ChEBI CHEBI:17203
IUPAC Name (2S)-pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

Melting Point ∽225°C (decomposition)
Beilstein 80810
Solubility Information Soluble in water and ethanol. Insoluble in ether,butanol and isopropanol.
Density 1.36
Quantity 100 g
Formula Weight 115.13
Sensitivity Hygroscopic
Chemical Name or Material L-Proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.