missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, 99%, Thermo Scientific Chemicals

Catalog Number 11459512
Click to view available options
Quantity:
25 g
100 g
500 g
147-85-3
C5H9NO2
115.13
MFCD00064318
ONIBWKKTOPOVIA-UHFFFAOYNA-N
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
145742
CHEBI:17203
(2S)-pyrrolidine-2-carboxylic acid
OC(=O)C1CCCN1
147-85-3
C5H9NO2
115.13
MFCD00064318
ONIBWKKTOPOVIA-UHFFFAOYNA-N
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
145742
CHEBI:17203
(2S)-pyrrolidine-2-carboxylic acid
OC(=O)C1CCCN1

L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in
TRUSTED_SUSTAINABILITY
Melting Point ∼225°C (decomposition)
Beilstein 80810
Merck Index 14,7780
Solubility Information Soluble in water and ethanol. Insoluble in ether,butanol and isopropanol.
Formula Weight 115.13
Percent Purity 99%
Density 1.36
Quantity 500 g
Optical Rotation −84° (c=1 in water)
Sensitivity Hygroscopic
Chemical Name or Material L-Proline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'