missing translation for 'onlineSavingsMsg'
Learn More

L-Prolinamide, 98%

Product Code. 10697864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697864 5 g Glass Bottle 5g
10227080 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10697864 Supplier Thermo Scientific Acros Supplier No. 302990050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7531-52-4 | C5H11N2O | 115.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7531-52-4
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005253
InChI Key VLJNHYLEOZPXFW-BYPYZUCNSA-O
Synonym l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
PubChem CID 111306
ChEBI CHEBI:21374
IUPAC Name (2S)-pyrrolidine-2-carboxamide
SMILES NC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 97.0°C to 102.0°C
Color Straw to White
Individual Impurity 97.5% min. (TLC)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation − 106.00
Specific Rotation Condition −106° (20°C c=2,EtOH)
Quantity 5 g
Formula Weight 114.15
Percent Purity 98%
Physical Form Powder or Granules
Chemical Name or Material L-Prolinamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.