Learn More
L-(-)-Prolinamide, 98%
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.
Solubility
Soluble in water and ethanol (50 mg/ml).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 95°C to 104°C |
| Quantity | 1 g |
| Beilstein | 80807 |
| Solubility Information | Soluble in water and ethanol (50mg/ml). |
| Optical Rotation | −105° (c=2 in Ethanol) |
| IUPAC Name | (2S)-pyrrolidine-2-carboxamide |
| Formula Weight | 114.15 |
| Percent Purity | 99% |
| Chemical Name or Material | L-(-)-Prolinamide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.