missing translation for 'onlineSavingsMsg'
Learn More

L-Praziquanamine, MedChemExpress

Product Code. 16459238
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459238 10 mg 10mg
16411658 5 mg 5mg
16403936 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16459238 Supplier Medchem Express Supplier No. HYN1765
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress L-Praziquanamine is a natural product.

L-Praziquanamine is a natural product.

Specifications

Chemical Name or Material L-Praziquanamine
CAS 99746-73-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H14N2O
Quantity 10 mg
Synonym (+)-Praziquanamine
Solubility Information DMSO : 100 mg/mL (494.44 mM; Need ultrasonic)
SMILES O=C1CNC[C@@]2([H])N1CCC3=C2C=CC=C3
Molecular Weight (g/mol) 202.25
Formula Weight 202.25
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.