missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L-Plastin Recombinant Rabbit Monoclonal Antibody (HL2501)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556777
This item is not returnable.
View return policy
Description
L-Plastin Recombinant Monoclonal Antibody for Western Blot, IHC
L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosterones in androgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.
Specifications
| L-Plastin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q6P698 | |
| LCP1 | |
| Synthetic peptide of zebrafish Lcp1. | |
| 100 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| HL2501 | |
| Unconjugated | |
| LCP1 | |
| 65 kDa macrophage protein; AW536232; bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); cb245; CP64; D14Ertd310e; DKFZp781A23186; epididymis secretory protein Li 37; fj24b12; FLJ25423; FLJ26114; FLJ39956; HEL-S-37; LC64P; LCP1; LCP-1; leucocyte-specific plastin 1; L-fimbrin; LPL; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (65 kDa macrophage protein) (PP65); L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); lymphocyte cytosolic plastin 1; lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein-1 (plasmin); plastin 2; plastin 2, L; plastin-2; Pls2; pp65; wu:fj24b12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 30583 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction