missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Pipecolinic acid, 99%, ACROS Organics™

Product Code. 10194840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194840 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10194840 Supplier Thermo Scientific Acros Supplier No. 218081000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3105-95-1
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H11NO2
MDL Number MFCD00005981
Quantity 100 mg
Beilstein 22, 8
Merck Index 14, 7458
Specific Rotation Condition −27.50° (24°C c=5,H2O)
Specific Rotation -24° to -27.5° (24°C, 589nm) (c=5, H2O)
Synonym l-pipecolic acid, s-piperidine-2-carboxylic acid, l-homoproline, h-hopro-oh, l-pipecolinic acid, l-pipecolate, 2s-piperidine-2-carboxylic acid, s-pipecolic acid, l--pipecolinic acid, l-piperidine-2-carboxylic acid
InChI Key HXEACLLIILLPRG-UHFFFAOYNA-N
SMILES OC(=O)C1CCCCN1
IUPAC Name piperidine-2-carboxylic acid
Molecular Weight (g/mol) 129.16
PubChem CID 439227
ChEBI CHEBI:30913
Formula Weight 129.16
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(-)-Pipecolinic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Pipecolinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.