missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, TRC

Product Code. 30132667
Change view
Click to view available options
Quantity:
1 g
25 g
50 g
Unit Size:
1g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132667 50 g 50g
30134213 25 g 25g
17959064 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132667 Supplier Toronto Research Chemicals Supplier No. TRCP31941550G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9 H11 N O2
Molecular Weight (g/mol) 165.19
Synonym L-Phenylalanine, Alanine, phenyl-, L- (4CI,7CI,8CI), (-)-Phenylalanine, (2S)-2-Amino-3-phenylpropanoic acid, (2s)-2-Amino-3-phenylpropanoic acid, (S)-(-)-Phenylalanine, (S)-2-Amino-3-phenylpropanoic acid, (S)-2-Amino-3-phenylpropionic acid, (S)-Phenylalanine, (S)-α-Amino-β-phenylpropionic acid, (S)-α-Aminobenzenepropanoic acid, (S)-α-Aminohydrocinnamic acid, 3-Phenyl-L-alanine, 3-Phenylalanine, Antibiotic FN 1636, Benzenepropanoic acid, α-amino-, (S)-, L-(-)-Phenylalanine, L-Alanine, 3-phenyl-, NSC 79477, Phenyl-α-alanine, Phenylalanine, β-Phenyl-L-alanine, β-Phenyl-α-alanine, Aspartame Imp. C (EP)
IUPAC Name (2S)-2-amino-3-phenylpropanoic acid
SMILES N[C@@H](Cc1ccccc1)C(=O)O

Specifications

InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1
Quantity 50 g
Formula Weight 165.079
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Phenylalanine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.