missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15577964 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15577964 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577964 Supplier Avantor J.T.Baker Supplier No. 208606

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63-91-2
Molecular Formula C9H11NO2
Packaging Poly Bottle
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES NC(CC1=CC=CC=C1)C(O)=O
IUPAC Name 2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
Quantity 1 kg
PubChem CID 6140
ChEBI CHEBI:17295
Formula Weight 165.19
Specific Gravity 1L = 1.29kg
Grade Multi-Compendial/USP
Chemical Name or Material L-Phenylalanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.