missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine Methyl Ester Hydrochloride, TRC

Product Code. 30416149
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416149 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416149 Supplier Toronto Research Chemicals Supplier No. TRCP31942610G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Phenylalanine Methyl Ester Hydrochloride
CAS 7524-50-7
Molecular Formula C10H13NO2.HCl
Quantity 10 g
Synonym L-Phenylalanine','methyl ester','hydrochloride (1:1), Alanine','phenyl-','methyl ester','hydrochloride','L- (7CI,8CI), L-Phenylalanine','methyl ester','hydrochloride (9CI), (2S)-2-Amino-3-phenylpropionic acid methyl ester hydrochloride, (S)-2-Amino-3-phenylpropionic acid methyl ester hydrochloride, (S)-Phenylalanine methyl ester hydrochloride, Methyl (2S)-2-amino-3-phenylpropanoate hydrochloride, Methyl (S)-2-amino-3-phenylpropanoate hydrochloride, Methyl (S)-phenylalaninate hydrochloride, Methyl L-phenylalaninate hydrochloride, NSC 194642, Phenylalanine hydrochloride methyl ester, Phenylalanine methyl ester hydrochloride
InChI Formula InChI=1S/C10H13NO2.ClH/c1-13-10(12)9(11)7-8-5-3-2-4-6-8;/h2-6,9H,7,11H2,1H3;1H/t9-;/m0./s1
SMILES Cl.COC(=O)[C@@H](N)Cc1ccccc1
IUPAC Name methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
Molecular Weight (g/mol) 215.677
Formula Weight 215.0713
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.