missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine methyl ester hydrochloride, 98%

Product Code. 11470323
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470323 50 g 50g
11460323 10 g 10g
11480323 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470323 Supplier Thermo Scientific Alfa Aesar Supplier No. A11455.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7524-50-7 | C10H14ClNO2 | 215.68 g/mol

Methyl L-phenylalaninate hydrochloride is used in pharmaceutical, food,animal feed,cosmetic,bio-chemical research etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl L-phenylalaninate hydrochloride is used in pharmaceutical, food,animal feed,cosmetic,bio-chemical research etc.

Solubility
It is soluble in methanol. (5mg/ ml-clear colorless solution)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents. Stable under recommended storage conditions. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7524-50-7
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00012489
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
Synonym l-phenylalanine methyl ester hydrochloride, h-phe-ome.hcl, methyl l-phenylalaninate hydrochloride, s-methyl 2-amino-3-phenylpropanoate hydrochloride, unii-47hk4y94ja, h-phe-ome hcl, h-phe-ome hydrochloride, l-phenylalanine, methyl ester, hydrochloride, l-phenylalanine methyl ester hcl
PubChem CID 75736
IUPAC Name methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1

Specifications

Melting Point 158°C to 162°C
Beilstein 3597948
Solubility Information It is soluble in methanol. (5mg/ ml-clear colorless solution)
Quantity 50 g
Formula Weight 215.68
Optical Rotation +37° (c=2 in ethanol)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Phenylalanine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.