missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine methyl ester hydrochloride, 98%

Product Code. p-181809 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10257163 2.5 g Glass bottle 2.5g
10367060 10 g Glass bottle 10g
10073473 50 g Glass bottle 50g
15109183 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10257163 Supplier Thermo Scientific Acros Supplier No. 130320025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7524-50-7 | C10H14ClNO2 | 215.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7524-50-7
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00012489
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
Synonym l-phenylalanine methyl ester hydrochloride, h-phe-ome.hcl, methyl l-phenylalaninate hydrochloride, s-methyl 2-amino-3-phenylpropanoate hydrochloride, unii-47hk4y94ja, h-phe-ome hcl, h-phe-ome hydrochloride, l-phenylalanine, methyl ester, hydrochloride, l-phenylalanine methyl ester hcl
PubChem CID 75736
SMILES [H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1

Specifications

Melting Point 156.0°C to 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5CH2CH(NH2)CO2CH3·HCl
Quantity 2.5 g
Beilstein 14,499
Specific Rotation Condition + 37.00 (20.00°C c=2,C2H5OH)
Specific Rotation + 37.00
IUPAC Name methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
Formula Weight 215.68
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material L-Phenylalanine methyl ester hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.