missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, MedChemExpress

Product Code. 16402519
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402519 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402519 Supplier Medchem Express Supplier No. HYN0215
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress L-Phenylalanine ((S)-2-Amino-3-phenylpropionic acid) is an essential amino acid isolated from Escherichia coli.

L-Phenylalanine is a α2δ subunit of voltage-dependent Ca+ channels antagonist with a Ki of 980 nM. L-phenylalanine is a competitive antagonist for the glycine- and glutamate-binding sites of N-methyl-D-aspartate receptors (NMDARs) (KB of 573 μM ) and non-NMDARs, respectively. L-Phenylalanine is widely used in the production of food flavors and pharmaceuticals.

Specifications

Chemical Name or Material L-Phenylalanine
Physical Form Solid
CAS 63-91-2
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C9H11NO2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym (S)-2-Amino-3-phenylpropionic acid
Solubility Information H2O : 6.67 mg/mL (40.38 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 165.19
Formula Weight 165.19
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.