missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine benzyl ester hydrochloride, 98%

Product Code. 11412040
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412040 25 g 25g
11402040 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412040 Supplier Thermo Scientific Alfa Aesar Supplier No. H28407.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2462-32-0 | C16H18ClNO2 | 291.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2462-32-0
Molecular Formula C16H18ClNO2
Molecular Weight (g/mol) 291.78
MDL Number MFCD00043249
InChI Key CEXFHIYDTRNBJD-RSAXXLAASA-N
Synonym l-phenylalanine benzyl ester hydrochloride, h-phe-obzl.hcl, h-phe-obzl hcl, l-phenylalanine benzyl ester hcl, s-benzyl 2-amino-3-phenylpropanoate hydrochloride, phenylalanine, phenylmethyl ester, hydrochloride, benzyl 2s-2-amino-3-phenylpropanoate hydrochloride, phenylmethyl 2s-2-amino-3-phenylpropanoate, chloride, pubchem19007, benzyl phenylalaninate hydrochloride
PubChem CID 12636208
IUPAC Name benzyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 197°C to 200°C
Quantity 25 g
Formula Weight 291.77
Percent Purity 98%
Chemical Name or Material L-Phenylalanine benzyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.