missing translation for 'onlineSavingsMsg'
Learn More

L-Penicillamine Hydrochloride, TRC

Product Code. 30428354
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428354 250 mg 250mg
30409557 1 g -
30424749 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428354 Supplier Toronto Research Chemicals Supplier No. TRCP223015250MG
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25526-04-9
Molecular Formula C5H11NO2S . (HCl)
Molecular Weight (g/mol) 185.67
Synonym 3-Mercapto-L-valine Hydrochloride, L-Penicillamine Hydrochloride, NSC 81988, (R)-2-Amino-3-mercapto-3-methylbutanoic Acid Hydrochloride
IUPAC Name (2R)-2-amino-3-methyl-3-sulfanylbutanoic acid;hydrochloride
SMILES Cl.CC(C)(S)[C@H](N)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H11NO2S.ClH/c1-5(2,9)3(6)4(7)8;/h3,9H,6H2,1-2H3,(H,7,8);1H/t3-;/m1./s1
Formula Weight 185.0277
Chemical Name or Material L-Penicillamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.