missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Penicillamine, 98%, Thermo Scientific Chemicals

Catalog Number 10279750
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1113-41-3
C5H11NO2S
149.21
MFCD00064303
VVNCNSJFMMFHPL-GSVOUGTGSA-N
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
92863
CHEBI:50869
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
CC(C)(C(C(=O)O)N)S
This item is not returnable. View return policy
1113-41-3
C5H11NO2S
149.21
MFCD00064303
VVNCNSJFMMFHPL-GSVOUGTGSA-N
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
92863
CHEBI:50869
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
CC(C)(C(C(=O)O)N)S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 206.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2C(SH)CH(NH2)CO2H
Quantity 1 g
Beilstein 04, III, 1662
Merck Index 15, 7197
Specific Rotation Condition +63° (20°C c=5,1 M NaOH)
Specific Rotation + 63.00
Solubility Information (5% in water) Clear colorless
Formula Weight 149.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Penicillamine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Penicillamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'