missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Pantolactone, 98%, Thermo Scientific™

Product Code. 15422178
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422178 250 mg 250mg
15412178 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422178 Supplier Thermo Scientific Alfa Aesar Supplier No. H55822.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-(+)-Pantolactone, is used as Pharmaceutical material and intermediate. It is used as an Intermediate in organic syntheses.

Solubility
Soluble in water, hardly soluble in alcohol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Heat sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5405-40-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00216625
InChI Key SERHXTVXHNVDKA-SCSAIBSYSA-N
Synonym s-+-pantolactone, l-pantolactone, s-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, s-pantolactone, l-+-pantoyl lactone, 3s-3-hydroxy-4,4-dimethyloxolan-2-one, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3s, pantolactons, l-pantoyl lactone
PubChem CID 736053
IUPAC Name (3S)-3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 89°C to 94°C
Boiling Point 120°C to 122°C (15 mmHg)
Quantity 250 mg
Beilstein 4659425
Solubility Information Soluble in water,hardly soluble in alcohol.
Optical Rotation +51° (c=3 in Water)
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material L-(+)-Pantolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.