missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine, MedChemExpress

Product Code. p-200101234
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16482266

Brand: Medchem Express HYB1352

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-ornithine has an antifatigue effect in increasing the efficiency of energy consumption and promoting the excretion of ammonia.

L-ornithine has an antifatigue effect in increasing the efficiency of energy consumption and promoting the excretion of ammonia.

Specifications

Chemical Name or Material L-Ornithine
CAS 70-26-8
Color White
Purity Grade Notes Research
Molecular Formula C5H12N2O2
Quantity 500 mg
Synonym (S)-2,5-Diaminopentanoic acid
Solubility Information H2O : 50 mg/mL (378.33 mM; Need ultrasonic) ∣DMSO : 1.3 mg/mL (9.84 mM; Need ultrasonic)
SMILES N[C@@H](CCCN)C(O)=O
Molecular Weight (g/mol) 132.16
Formula Weight 132.16
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.