missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine L-Aspartate Salt, TRC

Product Code. 30409243
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409243 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409243 Supplier Toronto Research Chemicals Supplier No. TRCO6888451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Ornithine L-Aspartate
CAS 3230-94-2
Purity Grade Notes HPLC
Molecular Formula C5 H12 N2 O2 . C4 H7 N O4
Quantity 1 g
Synonym L-Aspartic acid','compd. with L-ornithine (1:1), Aspartic acid','L-','compd. with L-ornithine (1:1) (8CI), L-Ornithine','L-aspartate (1:1), Ornithine','L-','L-aspartate (1:1) (8CI), Aspartic acid compd. with ornithine, Hepamerz, L-Ornithine L-aspartate, Ormeta, Ornithine aspartate, Orparan, Ruigan
InChI Formula InChI=1S/C5H12N2O2.C4H7NO4/c6-3-1-2-4(7)5(8)9;5-2(4(8)9)1-3(6)7/h4H,1-3,6-7H2,(H,8,9);2H,1,5H2,(H,6,7)(H,8,9)/t4-;2-/m00/s1
SMILES NCCC[C@H](N)C(=O)O.N[C@@H](CC(=O)O)C(=O)O
IUPAC Name (2S)-2-aminobutanedioic acid;(2S)-2,5-diaminopentanoic acid
Molecular Weight (g/mol) 265.26
Formula Weight 265.1274
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.