missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride MP Biomedicals

Product Code. 11493090 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493090 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493090 Supplier MP Biomedicals Supplier No. 0210042180
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Quantity 100 g
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
SMILES C(CC(C(=O)O)N)CN.Cl
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
Molecular Weight (g/mol) 168.621
PubChem CID 76654
Formula Weight 168.6
Chemical Name or Material L-Ornithine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.