missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride MP Biomedicals

Product Code. 11483090 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
1 kg
Unit Size:
1kg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483090 5 g 5g
11403100 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483090 Supplier MP Biomedicals Supplier No. 0210042105
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6153-39-5
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
InChI Key NBKPNAMTHBIMLA-UHFFFAOYSA-N
Synonym 5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate
PubChem CID 3083941
IUPAC Name 5-methylbenzene-1,3-diol;hydrate
SMILES CC1=CC(=CC(=C1)O)O.O

Specifications

Assay Percent Range ∽ 99%
Quantity 5 g
Formula Weight 168.6
Physical Form Powder
Chemical Name or Material L-Ornithine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.