missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine hydrochloride, MedChemExpress

Product Code. 16479386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479386 100 mg 100mg
16472506 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479386 Supplier Medchem Express Supplier No. HYW017018100mg
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Ornithine hydrochloride is a free amino acid that plays a central role in the urea cycle and is also important for the disposal of excess nitrogen.

L-Ornithine hydrochloride is a free amino acid that plays a central role in the urea cycle and is also important for the disposal of excess nitrogen.

Specifications

Chemical Name or Material L-Ornithine hydrochloride
CAS 3184-13-2
Color White
Purity Grade Notes Research
Molecular Formula C5H13ClN2O2
Quantity 100 mg
Solubility Information H2O : ≥ 100 mg/mL (593.05 mM) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES NCCC[C@@](N)([H])C(O)=O.Cl
Molecular Weight (g/mol) 168.62
Formula Weight 168.62
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.