missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ornithine hydrochloride, 99%

Product Code. 10259393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10259393 500 g Plastic bottle 500g
10572141 25 g Plastic bottle 25g
10666522 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10259393 Supplier Thermo Scientific Acros Supplier No. 129565000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3184-13-2 | C5H12N2O2·HCl | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H12N2O2·HCl
Molecular Weight (g/mol) 168.62
MDL Number MFCD00064562
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Melting Point 245.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula H2N(CH2)3CH(NH2)CO2H·HCl
Quantity 500 g
Beilstein 04, 420
Merck Index 14, 6874
Specific Rotation Condition + 24.00 (20.00°C c=4,6N HCl)
Specific Rotation + 24.00
Solubility Information (5% in 1 N HCl) Clear colorless
Formula Weight 168.62
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L(+)-Ornithine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.