missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals
14.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.
Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.
Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications
Melting Point | ∼238°C (decomposition) |
Beilstein | 3625847 |
Merck Index | 14,6874 |
Solubility Information | Soluble in water (543mg/ml at 20°C),and alcohol. |
Formula Weight | 168.62 |
Percent Purity | 99% |
Quantity | 25 g |
Optical Rotation | +23° (c=5 in 5N HCl) |
Chemical Name or Material | L-Ornithine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'