missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Ornithine hydrochloride

Product Code. 15423049
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423049 50 g 50g
15410127 100 g 100g
15420127 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15423049 Supplier Thermo Scientific Chemicals Supplier No. J60241.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3184-13-2 | C5H13ClN2O2 | 168.621 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
MDL Number MFCD00064562
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Melting Point ∽238°C (decomposition)
Color White
Beilstein 3625847
Merck Index 14,6874
Quantity 50 g
Formula Weight 168.62
Optical Rotation +23° (c=5 in 5N HCl)
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material L-Ornithine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.