missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford L-Ornithine HCL
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Description
Polyamine growth regulator.
Polyamine growth regulator.
Specifications
Specifications
| CAS | 3184-13-2 |
| Molecular Formula | C5H12N2O2-HCl |
| Quantity | 25 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction