missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L(+)-Norvaline, 99%
£27.40 - £694.00
Chemical Identifiers
CAS | 6600-40-4 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00064421 |
InChI Key | SNDPXSYFESPGGJ-BYPYZUCNSA-N |
Synonym | l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline |
PubChem CID | 65098 |
ChEBI | CHEBI:18314 |
IUPAC Name | (2S)-2-aminopentanoic acid |
SMILES | CCC[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10677552
|
Thermo Scientific Chemicals
226020010 |
1 g | Glass Bottle |
£27.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10299493
|
Thermo Scientific Chemicals
226020050 |
5 g | Glass Bottle |
£85.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10036193
|
Thermo Scientific Chemicals
226020500 |
50 g | Glass Bottle |
£694.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6600-40-4 | |
117.15 | |
SNDPXSYFESPGGJ-BYPYZUCNSA-N | |
65098 | |
(2S)-2-aminopentanoic acid |
C5H11NO2 | |
MFCD00064421 | |
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline | |
CHEBI:18314 | |
CCC[C@H](N)C(O)=O |
Specifications
6600-40-4 | |
99% | |
Authentic | |
04, 416 | |
(TLC) not detectable | |
SNDPXSYFESPGGJ-BYPYZUCNSA-N | |
(2S)-2-aminopentanoic acid | |
117.15 | |
CHEBI:18314 | |
98.5 to 101.5% | |
Fine Crystalline Powder |
White | |
C5H11NO2 | |
MFCD00064421 | |
15, 6805 | |
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline | |
CCC[C@H](N)C(O)=O | |
+23.5° to +25.5° (20°C, 589nm) (c=10, 6N HCl) | |
65098 | |
117.15 | |
24.50 (20.00°C c=10,6 N HCl) | |
L(+)-Norvaline |
RUO – Research Use Only