missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Norleucine, MedChemExpress
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
L-Norleucine ((S)-2-Aminohexanoic acid) is an isomer of leucine, specifically affects protein synthesis in skeletal muscle, and has antivirus activity.
Specifications
Specifications
| Chemical Name or Material | L-Norleucine |
| CAS | 327-57-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C6H13NO2 |
| Synonym | (S)-2-Aminohexanoic acid (S)-Norleucine |
| Solubility Information | H2O : 10 mg/mL (76.24 mM; Need ultrasonic) |
| SMILES | N[C@@H](CCCC)C(O)=O |
| Molecular Weight (g/mol) | 131.17 |
| Quantity | 1 g |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction