missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-NMMA acetate
GREENER_CHOICE

Product Code. 18005962
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18005962 10 mg 10mg
30001127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18005962 Supplier Tocris Bioscience™ Supplier No. 0771/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Non-selective NOS inhibitor

Competitive, irreversible inhibitor of all three NOS isoforms.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53308-83-1
Molecular Formula C9H20N4O4

Specifications

Quantity 10 mg
Formula Weight 248.28
Purity >98%
Molecular Weight (g/mol) 248.283
Chemical Name or Material L-NMMA acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.