missing translation for 'onlineSavingsMsg'
Learn More

L-NMMA acetate, MedChemExpress

Product Code. 16413616
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16413616

Brand: Medchem Express HY18732A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress L-NMMA acetate is a nitric oxide synthase inhibitor of all NOS isoforms including NOS1, NOS2, and NOS3. The Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are approximately 0.18, 0.4, and 6 μM, respectively.

L-NMMA acetate is a nitric oxide synthase inhibitor of all NOS isoforms including NOS1, NOS2, and NOS3. The Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are approximately 0.18, 0.4, and 6 μM, respectively.

Specifications

Chemical Name or Material L-NMMA acetate
Physical Form Solid
CAS 53308-83-1
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C9H20N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Tilarginine acetateMethylarginine acetate
Solubility Information H2O : ≥ 50 mg/mL (201.39 mM)
SMILES N[C@@H](CCCNC(NC)=N)C(O)=O.O=C(C)O
Molecular Weight (g/mol) 248.28
Formula Weight 248.28
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.