missing translation for 'onlineSavingsMsg'
Learn More

L-NMMA acetate, MedChemExpress

Product Code. 16407618
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16407618 10 mg 10mg
16483627 50 mg 50mg
16467526 100 mg 100mg
16413616 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16407618 Supplier Medchem Express Supplier No. HY18732A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-NMMA acetate is a nitric oxide synthase inhibitor of all NOS isoforms including NOS1, NOS2, and NOS3. The Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are approximately 0.18, 0.4, and 6 μM, respectively.

L-NMMA acetate is a nitric oxide synthase inhibitor of all NOS isoforms including NOS1, NOS2, and NOS3. The Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are approximately 0.18, 0.4, and 6 μM, respectively.

Specifications

Chemical Name or Material L-NMMA acetate
Physical Form Solid
CAS 53308-83-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C9H20N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Tilarginine acetateMethylarginine acetate
Solubility Information H2O : ≥ 50 mg/mL (201.39 mM)
SMILES N[C@@H](CCCNC(NC)=N)C(O)=O.O=C(C)O
Molecular Weight (g/mol) 248.28
Formula Weight 248.28
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.