missing translation for 'onlineSavingsMsg'
Learn More

L-NIO dihydrochloride, MedChemExpress

Product Code. 16483228
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16483228

Brand: Medchem Express HY100986

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-NIO dihydrochloride is a potent, non-selective and NADPH-dependent nitric oxide synthase (NOS) inhibitor, with Kis of 1.7, 3.9, 3.9 μM for neuronal (nNOS), endothelial (eNOS), and inducible (iNOS), respectively. L-NIO dihydrochloride induces a consistentfocal ischemic infarctin rats.

L-NIO dihydrochloride is a potent, non-selective and NADPH-dependent nitric oxide synthase (NOS) inhibitor, with Kis of 1.7, 3.9, 3.9 μM for neuronal (nNOS), endothelial (eNOS), and inducible (iNOS), respectively. L-NIO dihydrochloride induces a consistentfocal ischemic infarctin rats.

Specifications

Chemical Name or Material L-NIO dihydrochloride
Physical Form Oil
CAS 159190-44-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C7H17Cl2N3O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 125 mg/mL (507.86 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (135.42 mM; Need ultrasonic)
SMILES N[C@@H](CCCNC(C)=N)C(O)=O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 246.13
Formula Weight 246.13
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.