missing translation for 'onlineSavingsMsg'
Learn More

l-Nicotine, 1mg/ml in Methanol, Lipomed

Product Code. 30459391
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459391 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459391 Supplier Lipomed Supplier No. LPMNIC756FB1LM
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

l-Nicotine, 1mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotine
CAS 54-11-5
Analyte or Component Names Nicotine
Shipping Condition Room Temperature
Molecular Formula C10 H14 N2
Concentration 1 mg/mL
Synonym Nicotine, (-)-Nicotine, (-)-(S)-Nicotine, (-)-3-(1-Methyl-2-pyrrolidyl)pyridine, (S)-(-)-Nicotine, Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI), Nicotine (8 CI)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN1CCC[C@H]1c2cccnc2
IUPAC Name 3-[(2 S)-1-methylpyrrolidin-2-yl]pyridine
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 162.23
Quantity 1 mL
Formula Weight 162.1157 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.