missing translation for 'onlineSavingsMsg'
Learn More

L-NAME hydrochloride, MedChemExpress

Product Code. 16461896
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461896 500 mg 500mg
16447526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461896 Supplier Medchem Express Supplier No. HY18729A
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. L-NAME is a precursor to NOS inhibitor L-NOARG which has an IC50 value of 1.4 μM.

L-NAME hydrochloride inhibits NOS with an IC50 of 70 μM. L-NAME is a precursor to NOS inhibitor L-NOARG which has an IC50 value of 1.4 μM.

Specifications

Chemical Name or Material L-NAME hydrochloride
CAS 51298-62-5
Color White
Purity Grade Notes Research
Molecular Formula C7H16ClN5O4
Quantity 500 mg
Synonym NG-Nitroarginine methyl ester hydrochloride
Solubility Information H2O : 100 mg/mL (370.80 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (370.80 mM; Need ultrasonic)
SMILES N[C@@H](CCCNC(N[N+]([O-])=O)=N)C(OC)=O.[H]Cl
Molecular Weight (g/mol) 269.69
Formula Weight 269.69
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.