missing translation for 'onlineSavingsMsg'
Learn More

L-myo-Inositol-1,4,5-trisphosphate sodium salt, 98%, Thermo Scientific™

Product Code. 15458599
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458599 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458599 Supplier Thermo Scientific Alfa Aesar Supplier No. J67018.!Z
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2068-89-5
Color White
Molecular Formula C6H12Na3O15P3
MDL Number MFCD18427941
Quantity 0.1 mg
Synonym 1,4,5-ip3 sodium salt, l-myo-inositol-1,4,5-triphosphate sodium salt
InChI Key ZVCVTWVBDIPWPZ-FWMAINDVSA-K
SMILES C1(C(C(C(C(C1OP(=O)(O)[O-])O)OP(=O)(O)[O-])OP(=O)(O)[O-])O)O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;[(1S,2R,3S,4S,5R,6S)-2,3,5-trihydroxy-4,6-bis[[hydroxy(oxido)phosphoryl]oxy]cyclohexyl] hydrogen phosphate
Molecular Weight (g/mol) 486.038
PubChem CID 91886089
Formula Weight 486.04
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material L-myo-Inositol-1,4,5-trisphosphate sodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.