missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine sulfoximine, 98+%

Product Code. 10073130 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10073130 1 g Glass bottle 1g
10608134 250 mg Glass bottle 250mg
10607944 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10073130 Supplier Thermo Scientific Chemicals Supplier No. 227200010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15985-39-4 | C5H12N2O3S | 180.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15985-39-4
Molecular Formula C5H12N2O3S
Molecular Weight (g/mol) 180.22
MDL Number MFCD00002621
InChI Key SXTAYKAGBXMACB-DPVSGNNYSA-N
Synonym l-methionine sulfoximine, 2s-2-amino-4-s-methylsulfonimidoyl butanoic acid, l-methionine r,s-sulfoximine, methionine sulfoximine l, l-s-3-amino-3-carboxypropyl-s-methylsulfoximine, butanoic acid, 2-amino-4-s-methylsulfonimidoyl-, 2s, l-methionine-s,r-sulfoximine, prestwick_805, l-methionine-sulfoximine
PubChem CID 89034
ChEBI CHEBI:28490
IUPAC Name (2S)-2-amino-4-(methylsulfonimidoyl)butanoic acid
SMILES CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O

Specifications

Melting Point 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3S(O)(=NH)CH2CH2CH(NH2)CO2H
Quantity 1 g
Specific Rotation Condition +13.50° (20°C c=1,H2O)
Specific Rotation + 13.50
Solubility Information Solubility in water: soluble.
Formula Weight 180.22
Percent Purity 98+%
Physical Form Solid
Chemical Name or Material L-Methionine sulfoximine, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • L-Methionine sulfoximine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.