missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine Sulfoxide,13C, TRC

Product Code. 30138503
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138503 25 mg 25mg
30138395 10 mg 10mg
30003721 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138503 Supplier Toronto Research Chemicals Supplier No. TRCM26044825MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C C4 H11 N O3 S
Molecular Weight (g/mol) 166.203
IUPAC Name (2S)-2-amino-4-(113C)methylsulfinylbutanoic acid
SMILES [13CH3]S(=O)CC[C@H](N)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H11NO3S/c1-10(9)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-,10?/m0/s1/i1+1
Formula Weight 166.049
Chemical Name or Material L-Methionine Sulfoxide,13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.