missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine [R,S]-Sulfoximine, TRC

Product Code. 30138475
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138475 5 g 5g
30010349 1 g 1g
30011235 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138475 Supplier Toronto Research Chemicals Supplier No. TRCM2605005G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15985-39-4
Molecular Formula C5 H12 N2 O3 S
Molecular Weight (g/mol) 180.23
IUPAC Name (2S)-2-amino-4-(methylsulfonimidoyl)butanoic acid
SMILES CS(=N)(=O)CC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H12N2O3S/c1-11(7,10)3-2-4(6)5(8)9/h4,7H,2-3,6H2,1H3,(H,8,9)/t4-,11?/m0/s1
Quantity 5 g
Formula Weight 180.0569
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Methionine [R,S]-Sulfoximine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.