missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine methyl ester hydrochloride, 99%

Product Code. 11448506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448506 5 g 5g
11458506 25 g 25g
11468506 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448506 Supplier Thermo Scientific Alfa Aesar Supplier No. A15952.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2491-18-1 | C6H14ClNO2S | 199.69 g/mol

L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-18-1
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00012491,MFCD00067540
InChI Key MEVUPUNLVKELNV-UHFFFAOYNA-N
Synonym h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
PubChem CID 11435579
IUPAC Name methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(N)CCSC

Specifications

Melting Point 151°C to 152°C
Beilstein 3913214
Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 199.7
Optical Rotation +26° (c=5 in Water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Methionine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.