missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine methyl ester hydrochloride, 98%

Product Code. 10421851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
Packaging:
Glass bottle
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10421851 25 g Glass bottle 25g
10542441 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10421851 Supplier Thermo Scientific Chemicals Supplier No. 229350250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2491-18-1 | C6H14ClNO2S | 199.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-18-1
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00012491,MFCD00067540
InChI Key MEVUPUNLVKELNV-UHFFFAOYNA-N
Synonym h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
PubChem CID 11435579
IUPAC Name methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(N)CCSC

Specifications

Melting Point 151°C to 153°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass bottle
Linear Formula CH3SCH2CH2CH(NH2)CO2CH3·HCl
Quantity 25 g
Specific Rotation Condition +26° (24°C c=5,H2O)
Specific Rotation + 26
Solubility Information (5% in methanol) Clear colorless to light yellow
Formula Weight 199.7
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Methionine methyl ester hydrochloride, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.