missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine-d3, TRC

Product Code. 30476984
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476984 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476984 Supplier Toronto Research Chemicals Supplier No. TRCM260442100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methionine d3
CAS 13010-53-2
Purity Grade Notes HPLC
Molecular Formula C5 2H3 H8 N O2 S
Quantity 100 mg
Synonym L-Methionine-methyl-d3, Methionine-methyl-d3, L- (8CI), L-[Methyl-D3]-methionine, L-Methionine-d3, L-Methionine-D3 (methyl-D3), (2S)-2-Amino-4-(methyl-d3)sulfanylbutanoic acid, (S)-2-Amino-4-[(methyl-d3)thio]butanoic acid, S-(Methyl-d3)-L-homocysteine
InChI Formula InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1D3
SMILES [2H]C([2H])([2H])SCC[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-4-(trideuteriomethylsulfanyl)butanoic acid
Molecular Weight (g/mol) 152.23
Formula Weight 152.0699
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.