missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, Cell Culture Reagent

Product Code. 15494389
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
Product Code. Quantity unitSize
15494389 100 g 100g
15484389 50 g 50g
15404399 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 15494389

Brand: Thermo Scientific Chemicals J61904.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 284°C (decomposition)
Color White
Beilstein 1722294
Merck Index 14,5975
Solubility Information Soluble in water
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Quantity 100 g
Formula Weight 149.21
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Methionine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.