missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, 99+%, Powder, MP Biomedicals™

Product Code. 11423520 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423520 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423520 Supplier MP Biomedicals Supplier No. 0210229101
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amino acid derivative for laboratory and research applications.

Specifications

CAS 63-68-3
Color White
Assay Percent Range 99+%
Molecular Formula C5H11NO2S
MDL Number MFCD00063097 MFCD00801344
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
SMILES CSCC[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol) 149.21
Quantity 1 kg
PubChem CID 6137
ChEBI CHEBI:16643
Formula Weight 149.21
Physical Form Crystalline Powder
Chemical Name or Material L-Methionine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.