missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine-13C5, TRC

Product Code. 30465711
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465711 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465711 Supplier Toronto Research Chemicals Supplier No. TRCM2604441MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Methionine-13C5
CAS 202326-57-6
Molecular Formula 13C5 H11 N O2 S
Quantity 1 mg
Synonym L-Methionine-methyl,1,2,3,4-13C5, L-Methionine-13C5 (9CI), L-Methionine-13C5, (2S)-2-Amino-4-(methyl-13C)sulfanylbutanoic-1,2,3,4-13C4 acid, (S)-2-Amino-4-[(methyl-13C)thio]butanoic-1,2,3,4-13C4 acid, S-Methyl-L-homocysteine-13C5
InChI Formula InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1+1,2+1,3+1,4+1,5+1
SMILES [13CH3]S[13CH2][13CH2][13C@H](N)[13C](=O)O
IUPAC Name (2S)-2-amino-4-(1^{13}C)methylsulfanyl(1,2,3,4-^{13}C_{4})butanoic acid
Molecular Weight (g/mol) 154.175
Formula Weight 154.068
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.